missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pan-keratin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26411-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is a pan-keratin antibody.
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.Spécification
| pan-keratin | |
| Polyclonal | |
| Unconjugated | |
| KRT10 | |
| 58 kDa cytokeratin, CK 5, CK5, Cytokeratin 5, K5, keratin 5, KRT5, KRT5A, Type II keratin Kb5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3848, 3849, 3850, 3851, 3852, 3853, 3855, 3856, 3858, 3861, 3866, 3868, 3880 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02533, P02538, P04264, P05787, P08727, P08729, P08779, P12035, P13645, P13647, P19012, P19013, P35908 | |
| KRT1, KRT10, KRT14, KRT15, KRT16, KRT19, KRT2, KRT3, KRT4, KRT5, KRT6A, KRT7, KRT8 | |
| KRT5 Fusion Protein Ag24184 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu