missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAX7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PAX7 Polyclonal Antibody for Western Blot, IHC (P)
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene.
Spécification
| PAX7 | |
| Polyclonal | |
| Unconjugated | |
| PAX7 | |
| FLJ37460; HUP1; paired box 7; paired box gene 7; paired box homeotic gene 7; paired box protein Pax-7; paired box transcription factor PAX7; paired domain gene 7; PAX7; Pax-7; PAX7 transcriptional factor; PAX7A; PAX7B; RGD1564360; RMS2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 18509, 5081 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P23759, P47239 | |
| PAX7 | |
| KLH conjugated synthetic peptide between 410-437 amino acids from the C-terminal region of human PAX7. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Spot an opportunity for improvement?Share a Content Correction