Learn More
PBR Antibody (CL13358) [PerCP], Novus Biologicals™
Code produit 30289008 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30289008 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30289008 Fournisseur Novus Biologicals Code fournisseur NBP318567PCP
Mouse Monoclonal Antibody
PBR Monoclonal antibody specifically detects PBR in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Spécification
Antigène PBR
Applications Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL13358
Conjugué PerCP
Formule PBS
Alias de gène benzodiazepine peripheral binding site, BZRPDBI, IBP, MBRPBS, mDRC, Mitochondrial benzodiazepine receptor, PBRBPBS, PBRS, peripheral benzodiazepine receptor, Peripheral-type benzodiazepine receptor, pk18, PKBSbenzodiazapine receptor (peripheral), PTBR, translocator protein, translocator protein (18kDa)
Espèces hôtes Mouse
Immunogène This antibody was developed using a synthetic peptide derived from P30536, with the exact immunogen sequence remaining proprietary.
Méthode de purification Protein A purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Cancer
Primaire ou secondaire Primary
Identification génétique (Entrez) 706
Espèces cibles Human
Contenu et stockage Store at 4⪚C in the dark.
Forme Purified
Isotype IgG2b