Learn More
PCNA Antibody (PCNA/8303R), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-23683-100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PCNA Monoclonal antibody specifically detects PCNA in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Spécification
| PCNA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen | |
| Recombinant full-length human PCNA protein | |
| 100 μg | |
| Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers | |
| 5111 | |
| Store at 4°C. | |
| IgG κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| PCNA/8303R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |