Learn More
PCSK9 Monoclonal Antibody (15), Invitrogen™
Mouse Monoclonal Antibody
258.00€ - 450.00€
Spécification
| Antigène | PCSK9 |
|---|---|
| Clone | 15 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
17759753
|
Invitrogen
MA543397 |
50 μL |
258.00€
50µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
17799543
![]() |
Invitrogen
MA542994 |
100 μL |
450.00€
100µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Specificity: Human PCSK9
PCSK9 is a proprotein convertase belonging to the proteinase K subfamily of the secretory subtilase family. This protein is synthesized as a soluble zymogen that undergoes autocatalytic intramolecular processing in the endoplasmic reticulum. The protein may function as a proprotein convertase. The protein plays a role in cholesterol homeostasis and may have a role in the differentiation of cortical neurons.Spécification
| PCSK9 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| AI415265; AI747682; convertase subtilisin; convertase subtilisin/kexin type 9 preproprotein; FH3; HCHOLA3; hypercholesterolemia; LDLCQ1; NARC1; NARC-1; neural apoptosis regulated convertase 1; neural apoptosis-regulated convertase 1; OTTHUMP00000009833; PC9; PCSK9; Proprotein convertase 9; proprotein convertase PC9; proprotein convertase subtilisin/kexin type 9; PSEC0052; Subtilisin/kexin-like protease PC9 | |
| PCSK9 | |
| IgG1 | |
| Protein A | |
| Antibody |
| 15 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8NBP7 | |
| 255738 | |
| Recombinant Human PCSK9 Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| PCSK9 |