missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Antibody (826722) [DyLight 488], Novus Biologicals™
Tous les produits Bio Techne ProduitsDescription
PD-ECGF/Thymidine Phosphorylase Monoclonal antibody specifically detects PD-ECGF/Thymidine Phosphorylase in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigène | PD-ECGF/Thymidine Phosphorylase |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 826722 |
| Conjugué | DyLight 488 |
| Formule | 50mM Sodium Borate |
| Alias de gène | EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase |
| Espèces hôtes | Rat |
| Immunogène | S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse PD-ECGF/Thymidine Phosphorylase, Met1-Pro471, Accession # Q99N42 |
| Méthode de purification | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?