Learn More
PeproTech PD-L1 Fc Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Marque: PeproTech 500-P321-50UG
Description
AA Sequence of recombinant protein: FTVTVPKDLY VVEYGSNMTI ECKFPVEKQL DLAALIVYWE MEDKNIIQFV HGEEDLKVQH SSYRQRARLL KDQLSLGNAA LQITDVKLQD AGVYRCMISY GGADYKRITV KVNAPYNKIN QRILVVDPVT SEHELTCQAE GYPKAEVIWT SSDHQVLSGK TTTTNSKREE KLFNVTSTLR INTTTNEIFY CTFRRLDPEE NHTAELVIPE LPLAHPPNER GGPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHN HYTQKSLSLS PGK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PD-L1 Fc. Anti-Human PD-L1 Fc-specific antibody was purified by affinity chromatography employing an immobilized Human PD-L1 Fc matrix. Sandwich ELISA: To detect Human PD-L1 Fc by sandwich ELISA (using 100 μL/well), a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human PD-L1 Fc (500-P321BT) as a detection antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human PD-L1 Fc. Western Blot: To detect Human PD-L1 Fc by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible secondary reagents, the detection limit for Recombinant Human PD-L1 Fc is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
Spécification
| PD-L1 Fc | |
| Polyclonal | |
| Unconjugated | |
| Cd274 | |
| A530045L16Rik; B7 homolog 1; B7-H; B7h1; B7-H1; Cd274; CD274 antigen; CD274 molecule; PDCD1 ligand 1; Pdcd1l1; Pdcd1lg1; Pdl1; PD-L1; pdl-1; Programmed cell death 1 ligand 1; programmed death ligand 1; RGD1566211; sCD274; soluble CD274; soluble PD L1; sPD L1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29126 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q9NZQ7 | |
| Cd274 | |
| CHO cells-derived Recombinant Human PD-L1 Fc. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.