missing translation for 'onlineSavingsMsg'
Learn More

PDE7B Antibody, Novus Biologicals™

Code produit 18381092 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18381092 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381092 Supplier Novus Biologicals Supplier No. NBP312441

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDE7B Polyclonal antibody specifically detects PDE7B in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigène PDE7B
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Conjugué Unconjugated
Formule Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Alias de gène bA472E5.1, cAMP-specific 3'-5'-cyclic phosphodiesterase 7B, EC 3.1.4, EC 3.1.4.17, high-affinity cAMP-specific 3'-5'-cyclic phosphodiesterase, MGC88256, phosphodiesterase 7B, rolipram-insensitive phosphodiesterase type 7
Espèces hôtes Rabbit
Immunogène Synthetic cyclic peptide common to all PDE7B variants.
Méthode de purification Affinity-purified
Quantité 100 μg
État réglementaire RUO
Disciplines de recherche Signal Transduction, Vision
Primaire ou secondaire Primary
Identification génétique (Entrez) 27115
Espèces cibles Human, Mouse, Rat, Bovine
Contenu et stockage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Forme Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.