Learn More
PDE8A Antibody (EM-52), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-62213MFV450
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PDE8A Monoclonal antibody specifically detects PDE8A in Human samples. It is validated for Western Blot
Spécification
| Western Blot | |
| Human PDE8a | |
| Primary | |
| IgG1 κ |
| Mouse | |
| Lipid and Metabolism, Signal Transduction | |
| Store at 4C in the dark. |