missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDGF-B Antibody (PDGFB/3071), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-08646AF647
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PDGF-B Monoclonal antibody specifically detects PDGF-B in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Spécification
| PDGF-B | |
| Monoclonal | |
| Alexa Fluor 647 | |
| B chain, Becaplermin, FLJ12858, PDGF subunit B, PDGF2, PDGF-2, platelet-derived growth factor 2, Platelet-derived growth factor beta polypeptide, platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis)oncogene homolog), platelet-derived growth factor subunit B, platelet-derived growth factor, B chain, Platelet-derived growth factor, beta polypeptide (oncogene SIS), Proto-oncogene c-Sis, SSV | |
| Recombinant fragment of human PDGF-B protein (around aa 27-158) (exact sequence is proprietary) (Uniprot: P01127) | |
| 100 μg | |
| Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers | |
| 5155 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| PDGFB/3071 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu