missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDX-1/IPF1 Antibody (267712) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals FAB2419JF635
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PDX-1/IPF1 Monoclonal antibody specifically detects PDX-1/IPF1 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),Immunocytochemistry
Spécification
| PDX-1/IPF1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| Glucose-sensitive factor, IDX-1GSF, Insulin promoter factor 1, insulin promoter factor 1, homeodomain transcription factor, Insulin upstream factor 1, IPF1pancreas/duodenum homeobox protein 1, Islet/duodenum homeobox-1, MODY4IUF1, pancreatic and duodenal homeobox 1, pancreatic-duodenal homeobox factor 1, PDX-1IPF-1, somatostatin transcription factor 1, Somatostatin-transactivating factor 1, STF-1IUF-1 | |
| E. coli-derived recombinant human PDX-1, Ala91-Arg283, Accession # P52945 | |
| 0.1 mL | |
| Lipid and Metabolism, Stem Cell Markers | |
| 3651 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 267712 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur