Learn More
Invitrogen™ Pectate lyase 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5144267
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Pectate lyase 1 Polyclonal Antibody for Western Blot, ELISA
Has pectate lyase activity.
Spécification
| Pectate lyase 1 | |
| Polyclonal | |
| Unconjugated | |
| allergen Cup a 1; EC 4.2.2.2; Major pollen allergen Cup a 1; Pectate lyase 1 | |
| Recombinant Hesperocyparis arizonica Pectate lyase 1 protein (22-367AA). | |
| 100 μg | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Rabbit | |
| Protein G | |
| RUO | |
| Plant | |
| Antibody | |
| IgG |