Learn More
MedChemExpress Pembrolizumab
MedChemExpress Pembrolizumab (MK-3475) is a humanized IgG4 antibody inhibiting the programmed cell death 1 (PD-1) receptor, used in cancer immunotherapy.
Marque:  MedChemExpress HY-P9902
Encompass_Preferred
Code nomenclature Nacres: NA.46
Code produit. 16452278
  • 934.20€ / 5mg
Spécification
PD-1/PD-L1
Please store the product under the recommended conditions in the Certificate of Analysis.
Shipping with dry ice
5 mg
Inhibitory Antibodies