Learn More
Invitrogen™ PENK Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA130928
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
neuropeptide hormone; plays a role in the responses to stress and pain perception.
Specifications
| PENK | |
| Polyclonal | |
| Unconjugated | |
| PENK | |
| AI326464; ENK; enkephalin; enkephalin A; Leu-enkephalin; Met-enkephalin; Met-enkephalin-Arg-Gly-Leu; Met-enkephalin-Arg-Phe; Met-enkephalin-Arg-Ser-Leu; OGF; Opioid growth factor; PE; Penk; PENK(114-133); PENK(143-183); PENK(143-184); PENK(143-185); PENK(237-258); PENK(238-259); PENK(239-260); Penk1; Penk2; PENK-A; Penk-rs; PPA; preproenkephalin; preproenkephalin 1; preproenkephalin 2; proenkephalin; proenkephalin 1; proenkephalin 2; proenkephalin related sequence; proenkephalin-A; spermatogenic-specific proenkephalin; Synenkephalin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18619, 29237 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen) | |
| 0.26 mg/mL | |
| PBS with 0.05% sodium azide | |
| P04094, P22005 | |
| PENK | |
| CSYSKEVPEMEKRYGGFMRF Conjugated to KLH. | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction