missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHD4/HIF Prolyl Hydroxylase 4 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NB100-295G
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PHD4/HIF Prolyl Hydroxylase 4 Polyclonal specifically detects PHD4/HIF Prolyl Hydroxylase 4 in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| PHD4/HIF Prolyl Hydroxylase 4 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| NP_808808.1 | |
| P4HTM | |
| The epitope recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 502) of human PHD4/HIF Prolyl Hydroxylase 4 using the numbering given in entry NP_808808.1 (GeneID 54681). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Does not appear to work in Mouse. Rat reactivity reported in scientific literature (PMID: 17003483). | |
| Store at 4C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EC 1.14.11.-, EGLN4, HIFPH4, HIF-PH4, HIF-prolyl hydroxylase 4, HPH-4, hypoxia-inducible factor prolyl 4-hydroxylase, Hypoxia-inducible factor prolyl hydroxylase 4, P4H with transmembrane domain, P4H-TMFLJ20262, PH-4, PH4 hypoxia inducible factor prolyl 4 hydroxylase, PHD4, proline 4-hydroxylase, prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum), Prolyl hydroxlase domain-containing 4, transmembrane prolyl 4-hydroxylase | |
| Rabbit | |
| 50 kDa | |
| 0.1 mL | |
| Cancer, Hypoxia, Lipid and Metabolism | |
| 54681 | |
| Human, Rat, Mouse (Negative) | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu