missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phenytoin Monoclonal Antibody (PY23-302.1)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA110736
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA1-10736 detects Phenytoin. MA1-10736 has been successfully used in ELISA applications. The MA1-10736 immunogen is Phenytoin, conjugated to KLH.
Phenytoin sodium is a commonly used antiepileptic. Phenytoin (fen-i-TOE-in) acts to suppress the abnormal brain activity seen in seizure by reducing electrical conductance among brain cells by stabilizing the inactive state of voltage-gated sodium channels. Aside from seizures, it is an option in the treatment of trigeminal neuralgia in the event that carbamazepine is deemed inappropriate.
Spécification
| Phenytoin | |
| Monoclonal | |
| 0.9 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| ELISA | |
| PY23-302.1 | |
| Unconjugated | |
| Dilantin; Epanutin; Infatabs; Kapseals; Phenytek | |
| Phenytoin, conjugated to KLH. | |
| 500 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu