missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phosphatidylinositol 4,5-bisphosphate Monoclonal Antibody (2C11)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA3500
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA3-500 detects PIP2 from human tissues. MA3-500 has been successfully used in ELISA, Immunoprecipitation, Immunofluorescence, and Western blotting procedures. The MA3-500 antigen is liposomes containing synthetic di-palmitoyl ptdlns(4,5)P2.
Phosphatidylinositol 4,5-bisphosphate or PIP2 is an important second messenger for a number of signaling processes. When certain extracellular signals are received, Phosphilipase C cleaves phosphatidylinositol into DAG and PIP2. PIP2 has been shown to interact with actin structural proteins including profiling, gelsolin, and cofilin, and demonstrates regulatory function of actin polymerization. It has also been shown to regulate vesicular transport by interacting with proteins like ADP-ribosylation factor and phospholipase D. PIP2 has also been shown to significantly affect intracellular calcium concentrations, which also regulate a variety of activities.
Spécification
| Phosphatidylinositol 4,5-bisphosphate | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.05% sodium azide | |
| Mouse | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
| 2C11 | |
| Unconjugated | |
| C45H85O19P3; PIP2 | |
| Liposomes (prepared in PBS with lipid A, phosphatidylcholine (PC) and cholesterol) containing synthetic di-palmitoyl PtdIns(4,5)P2. | |
| RUO | |
| Chemical | |
| Antibody | |
| IgM |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu