missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-FGFR3 (Tyr724) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA564798
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Phospho-FGFR3 (Tyr724) Polyclonal Antibody detects endogenous levels of FGFR3 only when phosphorylated at Tyr724.
FGFR3 is a member of the FGFR family of receptor tyrosine kinases. This family is known to regulate a host of cellular functions including angiogenesis, mitogenesis, osteogenesis, myogenesis, carcinogenesis, cellular differentiation, and tissue repair after injury. The FGFR family has also been implicated in a number of diseases including cancer, rheumatoid arthritis, and diabetic retinopathy. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein would consist of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds acidic and basic fibroblast growth hormone and plays a role in bone development and maintenance. Mutations in this gene lead to craniosynostosis and multiple types of skeletal dysplasia. Three alternatively spliced transcript variants that encode different protein isoforms have been described.
Spécification
| Phospho-FGFR3 (Tyr724) | |
| Polyclonal | |
| Unconjugated | |
| FGFR3 | |
| ACH; CD333; CEK2; FGF Receptor; FGFR; FGFR3; FGFR-3; FGFR3deltaTM; Fibroblast growth factor receptor 3; fibroblast growth factor receptor 3 variant 4; Flg-2; FR3; HBGFR; heparin-binding growth factor receptor; HSFGFR3EX; hydroxyaryl-protein kinase; JTK4; Mfr3; sam3; tyrosine kinase JTK4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14184, 2261, 84489 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P22607, Q61851 | |
| FGFR3 | |
| A synthesized peptide derived from human FGFR3 (Phospho-Tyr724). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu