missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-LCK (Tyr505) Monoclonal Antibody (SRRCHA), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
404.00€
Spécification
| Antigène | Phospho-LCK (Tyr505) |
|---|---|
| Clone | SRRCHA |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
This SRRCHA monoclonal antibody recognizes human and mouse LCK when phosphorylated on tyrosine 505 (Y505). Y505 is a negative-regulatory site that is phosphorylated by the kinase CSK. LCK is expressed in T cells where it associates with the cytoplasmic ta
LCK is a member of the Src-family tyrosine kinase, which are essential for signaling through the T-cell receptor (TCR) complex. Upon TCR triggering, LCK phosphorylates the ITAM motives in its zeta subunits establishing binding sites for the SH2 domains of the tyrosine kinase ZAP70, which is also phosphorylated by LCK and activated to generate subsequent signaling platforms by phosphorylation of adaptor LAT. The majority of LCK is localized to the plasma membrane, however, there is also a significant fraction associated with the Golgi apparatus which may contribute to Raf activation under conditions of weak stimulation through the TCR. LCK contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. LCK is also involved in the regulation of apoptosis induced by various stimuli, but not by the death receptors. Diseases associated with LCK include Immunodeficiency 22 and CD45 deficiency. Alternatively splice variants of the LCK gene encoding different isoforms of the protein have been found.Spécification
| Phospho-LCK (Tyr505) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P06239, P06240 | |
| LCK | |
| Primary | |
| Antibody |
| SRRCHA | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human, Mouse | |
| 16818, 3932 | |
| IgG1 κ | |
| 4°C, store in dark, DO NOT FREEZE! | |
| LCK |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit