Learn More
Phospho-MEK1 (Thr292) Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 28930-1-AP-20UL
Code nomenclature Nacres: NA.46
Code produit. 17291416
  • 135.00€ / 20µL
Description
MEK1 is also named as MAP2K1 (mitogen-activated protein kinase kinase 1), MAPKK1, PRKMK1, MKK1 and belongs to the MAP kinase kinase subfamily. It is a dual-specificity kinase that mediate ERK1 and ERK2 activation during adhesion and growth factor signaling(PMID:19219045). It also plays an essential role in extra-embryonic ectoderm during placentogenesis. MEK1 has 2 isoforms produced by alternative splicing with the molecular weight of 43 kDa and 41 kDa. MEK1 may form a heterodimer with Mek2.(PMID:21144847,19219045).
Spécification
Spécification
Phospho-MEK1 (Thr292)
Polyclonal
Unconjugated
20 μL
5604
Liquid
Western Blot, ELISA
450 μg/mL