missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-MYL9 (Thr19, Ser20) Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5105753
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody detects endogenous levels of MRLC1 only when phosphorylated at Thr19+Ser20.
Myosin, a structural component of muscle, consists of two heavy chains and four light chains. Myosin light chain may regulate muscle contraction by modulating the ATPase activity of myosin heads. It also binds calcium and is activated by myosin light chain kinase. Two transcript variants encoding different isoforms have been found.
Spécification
| Phospho-MYL9 (Thr19, Ser20) | |
| Polyclonal | |
| Unconjugated | |
| MYL9 | |
| 20 kDa myosin light chain; AI327049; DTNB; G1; Isoform L20-A; LC20; light polypeptide 9; MGC3505; MLC2; MLC-2; MLC20; MLC-2C; MRLC1; myl9; myl9.L; myl9b; myl9l; Mylc2c; myosin light chain 2; myosin light chain 9; myosin light chain 9 L homeolog; myosin light chain, regulatory C; myosin regulatory light chain; myosin regulatory light chain 1; myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2, smooth muscle major isoform; myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; myosin regulatory light polypeptide 9; myosin regulatory light polypeptide 9b; myosin RLC; myosin, light chain 9, regulatory; myosin, light chain 9b, regulatory; myosin, light polypeptide 9, regulatory; Myrl2; RLC-C; smooth muscle major isoform; unnamed protein product; XELAEV_18003093mg; zgc:77916 | |
| Rabbit | |
| Sequential chromatography | |
| RUO | |
| 10398, 296313, 98932 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P24844, Q64122, Q9CQ19 | |
| MYL9 | |
| A synthesized peptide derived from human MYL9(Accession P24844), corresponding to amino acid residues around phosphorylated Ser20. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu