Learn More
Phospho-Tyrosine Alkaline Phosphatase-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marque: R&D Systems APM1676
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Phospho-Tyrosine Monoclonal specifically detects Phospho-Tyrosine in Multi-Species samples. It is validated for Western Blot, ELISA.
Spécification
| Phospho-Tyrosine | |
| Monoclonal | |
| Alkaline Phosphatase (AP) | |
| Supplied as a 0.2 μm filtered solution in phosphate-buffered saline (PBS) with stabilizers and preservatives. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects proteins containing phosphorylated Tyrosine residues. ELISA and 2D Western blot analyses using pervanadate-treated cell lysates indicate that it binds Phospho-Tyrosine in a broad manner largely independent of the surrounding amino acid sequence. This antibody does not cross-react with proteins or peptides containing phosphorylated serine or threonine residues. | |
| Multi-species | |
| Purified |
| Western Blot, ELISA | |
| 179003 | |
| Western Blot 1:1000 dilution, ELISA | |
| 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, PhosphoTyrosine, pTyrosine, Tyrosine | |
| KLH-coupled Phospho-Tyrosine synthetic peptide. | |
| 100 μL | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. | |
| IgG1 |