missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIEZO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5143835
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PIEZO2 Polyclonal Antibody for Western Blot
Component of mechanosensitive channel required for rapidly adapting mechanically activated (MA) currents. Requires for Merkel-cell mechanotransduction.
Spécification
| PIEZO2 | |
| Polyclonal | |
| Unconjugated | |
| PIEZO2 | |
| 5930434P17; 9030411M15Rik; 9430028L06Rik; C18orf30; C18orf58; DA3; DA5; Fam38b; FAM38B2; family with sequence similarity 38, member A pseudogene; family with sequence similarity 38, member B; family with sequence similarity 38, member B2; HsT748; HsT771; LOW QUALITY PROTEIN: piezo-type mechanosensitive ion channel component 2; MWKS; piezo type mechanosensitive ion channel component 2; Piezo2; piezo-type mechanosensitive ion channel component 1 pseudogene; piezo-type mechanosensitive ion channel component 2; piezo-type mechanosensitive ion channel component 2 pseudogene; Protein FAM38B; protein PIEZO2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 63895, 667742 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q8CD54, Q9H5I5 | |
| PIEZO2 | |
| A peptide corresponding to 19 amino acids near the amino terminus of human PIEZO2. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu