Learn More
Novus Biologicals™ PIG3 Antibody (10A2), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-30804-25ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PIG3 Monoclonal antibody specifically detects PIG3 in Human samples. It is validated for Western Blot
Spécification
| PIG3 | |
| Monoclonal | |
| Unconjugated | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 10A2 | |
| Western Blot : 5-10 μg/mL | |
| p53-induced gene 3 protein, PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, tumor protein p53 inducible protein 3, Tumor protein p53-inducible protein 3 | |
| Hybridoma produced by the fusion of splenocytes from mice immunized with recombinant human PIG3 protein. | |
| 25 μg | |
| Apoptosis | |
| 9540 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |