missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 326.00€
Spécification
| Antigène | PIP |
|---|---|
| Dilution | Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
18626482
|
Novus Biologicals
NBP2-98684-100ul |
100 μg |
326.00€
100µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
18669611
|
Novus Biologicals
NBP2-98684-50ul |
50 μg |
216.00€
50µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
PIP Polyclonal antibody specifically detects PIP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Spécification
| PIP | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 5304 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PIP. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title