missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PKIA Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA538638
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PKIA Polyclonal Antibody for ICC/IF, IHC (P)
The protein encoded by this gene is a member of the cAMP-dependent protein kinase (PKA) inhibitor family. This protein was demonstrated to interact with and inhibit the activities of both C alpha and C beta catalytic subunits of the PKA. Alternatively spliced transcript variants encoding the same protein have been reported.
Spécification
| PKIA | |
| Polyclonal | |
| Unconjugated | |
| PKIA | |
| AI415001; cAMP-dependent protein kinase inhibitor alpha; cAMP-dependent protein kinase inhibitor, muscle/brain isoform; PKIA; PKIalpha; PKI-alpha; PRKACN1; protein kinase (cAMP-dependent, catalytic) inhibitor alpha; protein kinase inhibitor, alpha | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114906, 18767, 5569 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61925, P63248, P63249 | |
| PKIA | |
| A synthetic peptide derived from the internal region of human IPKA. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu