missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK1 Antibody (CL8360) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-21166F
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PLK1 Monoclonal antibody specifically detects PLK1 in Human samples. It is validated for Western Blot
Spécification
| PLK1 | |
| Monoclonal | |
| FITC | |
| cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13 | |
| This antibody was developed using a synthetic peptide derived from P53350, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers | |
| 5347 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot | |
| CL8360 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu