missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-30592
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PLS1 Polyclonal specifically detects PLS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| PLS1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Q14651 | |
| PLS1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DLIDAIAPNAVRQEMIRRENLSDEDKL | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| fimbrin, I plastin, intestine specific plastin, Intestine-specific plastin, I-plastin, plastin 1, plastin 1 (I isoform), Plastin-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 5357 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu