Learn More
PLXDC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA520466
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
A suggested positive control is human brain tissue lysate. PA5-20466 can be used with blocking peptide PEP-0583.
May play a role in tumor angiogenesis.
Spécification
| PLXDC2 | |
| Polyclonal | |
| Unconjugated | |
| PLXDC2 | |
| 1200007L24Rik; 5430431D22Rik; AI646728; AU022916; AV231634; plexin domain containing 2; plexin domain-containing protein 2; PLXDC2; TEM7R; tumor endothelial marker 7 related; tumor endothelial marker 7-related protein; UNQ2514/PRO6003 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 361282, 67448, 84898 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6UX71, Q9DC11 | |
| PLXDC2 | |
| An 18 amino acid peptide from near the amino terminus of human Plxdc2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |