missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PMPCB Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5100108
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody detects endogenous levels of total PMPCB.
PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.
Spécification
| PMPCB | |
| Polyclonal | |
| Unconjugated | |
| PMPCB | |
| 3110004O18Rik; Beta-MPP; mitochondrial processing peptidase beta subunit; mitochondrial-processing peptidase subunit beta; MPP11; MPPB; MPPP52; P-52; peptidase (mitochondrial processing) beta; peptidase, mitochondrial processing beta subunit; PMPCB | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64198, 73078, 9512 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75439, Q03346, Q9CXT8 | |
| PMPCB | |
| A synthesized peptide derived from human PMPCB(Accession O75439), corresponding to amino acid residues V303-F353. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu