missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ POLE4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA564473
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: GQEAIFILAR AAELFVETIA KDAYCCAQQG KRKTLQRRDL DNAIEAVDEF AFLEGTLD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
POLE4 is a histone-fold protein that interacts with other histone-fold proteins to bind DNA in a sequence-independent manner. These histone-fold protein dimers combine within larger enzymatic complexes for DNA transcription, replication, and packaging.
Spécification
| POLE4 | |
| Polyclonal | |
| Unconjugated | |
| POLE4 | |
| 2400007P05Rik; 5830430F06Rik; DNA polymerase epsilon 4, accessory subunit; DNA polymerase epsilon p12 subunit; DNA polymerase epsilon subunit 4; DNA polymerase epsilon subunit p12; DNA polymerase II subunit 4; DNA-directed DNA polymerase epsilon 4; p12; Pole4; polymerase (DNA) epsilon 4, accessory subunit; polymerase (DNA-directed), epsilon 4 (p12 subunit); polymerase (DNA-directed), epsilon 4, accessory subunit; YHHQ1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56655 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NR33 | |
| POLE4 | |
| Recombinant protein corresponding to Human POLE4. Recombinant protein control fragment (Product #RP-104126). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu