Learn More
PORCN Antibody, R&D Systems™
Code produit 18621509 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
25 μL
100 μL
Conditionnement:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18621509 25 μL 25µL
18285372 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18621509 Fournisseur Novus Biologicals Code fournisseur NBP25850425ul
 Article restreint
Expédition estimée: 01-12-2026
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
PORCN Polyclonal specifically detects PORCN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Spécification
Antigène PORCN
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Dilution Immunohistochemistry-Paraffin 1:50 - 1:200
Formule PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide
Alias de gène 2410004O13Rik, DHOF, EC 2.3.1.-, FODH, MG61PPNprobable protein-cysteine N-palmitoyltransferase porcupine, por, PORCMGC29687, porcupine homolog (Drosophila), Protein MG61
Symboles de gène(s) PORCN
Espèces hôtes Rabbit
Immunogène This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AVSFHFSNYFVGFLSEATATLAGAGFTEEKDHLEWDLTVSKPLNVELPRSMVEVVTSWNLPMSYWLNNYVFKNAL
Méthode de purification Affinity Purified
Quantité 25 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 64840
Spécificité du test Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Type de produit Antibody
Isotype IgG
Gamme de produits Novus
Afficher plus Afficher moins
For Research Use Only