Learn More
PREB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marque:  Novus Biologicals NBP1-87056
Code nomenclature Nacres: NA.46
Code produit. 18710984
  • 572.00€ / 0.10mL
Description
PREB Polyclonal specifically detects PREB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
For Research Use Only