missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRELP Monoclonal Antibody (01)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA530783
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human PRELP/Prolargin.
PRELP is a leucine-rich repeat protein present in connective tissue extracellular matrix. This protein functions as a molecule anchoring basement membranes to the underlying connective tissue. This protein has been shown to bind type I collagen to basement membranes and type II collagen to cartilage. It also binds the basement membrane heparan sulfate proteoglycan perlecan. This protein is suggested to be involved in the pathogenesis of Hutchinson-Gilford progeria (HGP), which is reported to lack the binding of collagen in basement membranes and cartilage.
Spécification
| PRELP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P51888 | |
| PRELP | |
| Recombinant Human PRELP/Prolargin Protein (Gln21-Ile382). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA | |
| 01 | |
| Unconjugated | |
| PRELP | |
| 55 kDa leucine-rich repeat protein of articular cartilage; 7330409J17Rik; MST161; MSTP161; PRELP; Prolargin; prolargin proteoglycan; proline and arginine rich end leucine rich repeat protein; proline arginine-rich end leucine-rich repeat; proline/arginine-rich end leucine-rich repeat protein; proline-arginine-rich end leucine-rich repeat protein; SLRR2A | |
| Mouse | |
| Protein A | |
| RUO | |
| 5549 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu