Learn More
Invitrogen™ Human PRKACA (PKA), His Tag Recombinant Protein
Human PRKACA (PKA), His Tag Recombinant Protein
Marque: Invitrogen™ P2912
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 3.18000kg
Description
PRKACA (PKA) is a serine⁄threonine protein kinase that phosphorylates a broad spectrum of substrates. PRKACA regulates many cellular processes including cell growth, cell differentiation, ion-channel conductivity, gene transcription, and synaptic release of neurotransmitters.Spécification
NP_002721.1 | |
5566 | |
10μg | |
PKA Kinase Family | |
Human | |
His Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
PRKACA | |
E. coli | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
43.5 kDa | |
PRKACA (PKA) | |
aa 1-351 | |
Recombinant | |
Storage Buffer: 30mM KPO4, pH 7.5, 50% glycerol, 150mM KCl, 1mM EDTA, 1mM DTT Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: AGC Group | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
protein kinase, cAMP-dependent, catalytic, alpha | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.