missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRKD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12785-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role. The protein encoded by this gene is one of the PKC family members. This kinase can be activated rapidly by the agonists of G protein-coupled receptors. It resides in both cytoplasm and nucleus, and its nuclear accumulation is found to be dramatically enhanced in response to its activation. This kinase can also be activated after B-cell antigen receptor (BCR) engagement, which requires intact phopholipase C gamma and the involvement of other PKC family members.Spécification
| PRKD3 | |
| Polyclonal | |
| Unconjugated | |
| PRKD3 | |
| EPK2, PKC NU, PKD3, PRKCN, PRKD3, Protein kinase C nu type, protein kinase D3, Protein kinase EPK2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23683, 313834, 75292 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O94806, Q8K1Y2 | |
| Prkd3 | |
| PRKD3 Fusion Protein Ag3499 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu