missing translation for 'onlineSavingsMsg'
Learn More

Progranulin/PGRN Antibody (007), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18466466 Shop All Bio Techne Products
Change view
Click to view available options
Quantité:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18466466 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18466466 Supplier Novus Biologicals Supplier No. NBP289675MFV610

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Progranulin/PGRN Monoclonal antibody specifically detects Progranulin/PGRN in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigène Progranulin/PGRN
Applications ELISA
Classification Monoclonal
Clone 007
Conjugué mFluor Violet 610 SE
Dilution ELISA
Formule 50mM Sodium Borate
Alias de gène CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN
Espèces hôtes Rabbit
Immunogène This antibody was obtained from a rabbit immunized with purified, recombinant Human Progranulin/PGRN (Accession#: NP_002078.1; Met 1-Leu 593).
Méthode de purification Protein A purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Cancer, Lipid and Metabolism
Primaire ou secondaire Primary
Identification génétique (Entrez) 2896
Espèces cibles Human
Contenu et stockage Store at 4C in the dark.
Forme Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.