missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Propamocarb Monoclonal Antibody (2H10)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA549660
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody tested in indirect ELISA.
Propamocarb is a systemic fungicide used to control a variety of fungal diseases in crops such as vegetables, fruits, and ornamental plants. It works by inhibiting the growth and reproduction of fungi, preventing them from causing further damage to the plants. Propamocarb is effective against a wide range of fungal pathogens, including Pythium and Phytophthora species.
Spécification
| Propamocarb | |
| Monoclonal | |
| 2.6 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of PPMC hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 2H10 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu