missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSF1 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-30366F
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PSF1 Polyclonal antibody specifically detects PSF1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)
Spécification
| PSF1 | |
| Polyclonal | |
| Immunoprecipitation, Western Blot (Negative) | |
| GINS complex subunit 1, GINS complex subunit 1 (Psf1 homolog), KIAA0186PSF1DNA replication complex GINS protein PSF1, partner of sld five-1 | |
| Maps to a region between residue 50 to 100 of human GINS Complex Subunit 1 using the numbering given in entry NP_066545.3 | |
| 0.1 mL | |
| Stem Cell Markers | |
| 9837 | |
| Store at 4C in the dark. | |
| IgG |
| Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction