missing translation for 'onlineSavingsMsg'
Learn More

PYHIN1 Antibody, Novus Biologicals™

Code produit 18280656 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18280656 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18280656 Supplier Novus Biologicals Supplier No. NBP179436

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PYHIN1 Polyclonal specifically detects PYHIN1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigène PYHIN1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugué Unconjugated
Dilution Western Blot 1.0 ug/ml
Formule PBS, 2% Sucrose with 0.09% Sodium Azide
Numéro d’ordre du gène NP_945148
Alias de gène IFIXMGC23885, Interferon-inducible protein X, pyrin and HIN domain family, member 1, pyrin and HIN domain-containing protein 1, RP11-520H16.1
Symboles de gène(s) PYHIN1
Espèces hôtes Rabbit
Immunogène Synthetic peptide directed towards the N terminal of human PYHIN1The immunogen for this antibody is PYHIN1. Peptide sequence KMKEEYDKIQIADLMEEKFPGDAGLGKLIEFFKEIPTLGDLAETLKREKL.
Poids moléculaire de l’antigène 51 kDa
Méthode de purification Affinity purified
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 149628
Spécificité du test Expected identity based on immunogen sequence: Human: 100%; Sumatran orangutan: 78%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.