missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ QARS Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA557902
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: HTGYVIELQH VVKGPSGCVE SLEVTCRRAD AGEKPKAFIH WVSQPLMCEV RLYERLFQHK NPEDPTEVPG GFLSDLNLAS LHVVDA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 100%.
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. In metazoans, 9 aminoacyl-tRNA synthetases specific for glutamine (gln), glutamic acid (glu), and 7 other amino acids are associated within a multienzyme complex. Although present in eukaryotes, glutaminyl-tRNA synthetase (QARS) is absent from many prokaryotes, mitochondria, and chloroplasts, in which Gln-tRNA(Gln) is formed by transamidation of the misacylated Glu-tRNA(Gln). Glutaminyl-tRNA synthetase belongs to the class-I aminoacyl-tRNA synthetase family.
Spécification
| QARS | |
| Polyclonal | |
| Unconjugated | |
| QARS1 | |
| 1110018N24Rik; 1200016L19Rik; C80286; GLNRS; glutamine-tRNA ligase; Glutamine--tRNA ligase; glutamine-tRNA synthetase; glutaminyl-tRNA synthetase; MSCCA; PRO2195; QARS; RGD1562301 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 290868, 5859, 97541 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P47897, Q66H61, Q8BML9 | |
| QARS1 | |
| Recombinant protein corresponding to Human QARS. Recombinant protein control fragment (Product #RP-96456). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu