missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB27B Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™
Tous les produits Bio Techne ProduitsDescription
RAB27B Polyclonal antibody specifically detects RAB27B in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigène | RAB27B |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugué | mFluor Violet 610 SE |
| Formule | 50mM Sodium Borate |
| Alias de gène | C25KG, RAB27B, member RAS oncogene family, ras-related protein Rab-27B |
| Espèces hôtes | Rabbit |
| Immunogène | Recombinant fusion protein containing a sequence corresponding to amino acids 1-218 of human RAB27B (NP_004154.2).,, Sequence:, MTDGDYDYLIKLLALGDSGVGKTTFLYRYTDNKFNPKFITTVGIDFREKRVVYNAQGPNGSSGKAFKVHLQLWDTAGQERFRSLTTAFFRDAMGFLLMFDLTSQQSFLNVRNWMSQLQANAYCENPDIVLIGNKADLPDQREVNERQARELADKYGIPYFETSAATGQNVEKAVETLLDLIMKRMEQCVEKTQIPDTVNGGNSGNLDGEKPPEKKCIC |
| Méthode de purification | Affinity purified |
| Quantité | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?