missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aqp3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Rat Aqp3.
Marque: Abnova PAB28874.100ug
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Sequence: CHLEQPPPSTEAENVKLAHMKHKEQISpécification
| Aqp3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant Rat Aqp3. | |
| PBS pH 7.4, 50% glycerol and 0.09% sodium azide | |
| NP_113891.1 | |
| Aqp3 | |
| Recombinant protein corresponding to Rat Aqp3 C-terminus. | |
| 100 μg | |
| Primary | |
| 0.5 ug/mL of PAB28874 was sufficient for detection of Aqp3, ∼31.5kDa and somtimes larger glycosylated bands ∼35-50kDa, in 10ug of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| Immunofluorescence (1:200-400) Western Blot (1:1000-4000) The optimal working dilution should be determined by the end user. | |
| Aqp3 | |
| MGC156623 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 65133 | |
| Human, Mouse, Rat | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu