Learn More
SSR1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SSR1.
Marque: Abnova PAB3091.400uL
Description
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34kDa glycoprotein encoded by this gene and a 22kDa glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are mostly noncanonical. [provided by RefSeq
Spécification
| SSR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SSR1. | |
| In PBS (0.09% sodium azide) | |
| DKFZp781N23103/FLJ14232/FLJ22100/FLJ23034/FLJ78242/FLJ93042/TRAPA | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6745 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| SSR1 | |
| SSR1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human SSR1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only