Learn More
TAAR1 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TAAR1.
Marque: Abnova PAB26396.50ug
Description
TAAR1 is a G protein-coupled receptor activated by trace amines. Trace amines are endogenous amine compounds that account for less than 1% of the biogenic amines in most brain regions (Bunzow et al., 2001 [PubMed 11723224]).[supplied by OMIM
Spécification
| TAAR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TAAR1. | |
| In PBS (0.09% sodium azide) | |
| MGC126874/MGC138399/RP11-295F4.9/TA1/TAR1/TRAR1 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 134864 | |
| Human, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6-15 ug/mL) The optimal working dilution should be determined by the end user. | |
| TAAR1 | |
| TAAR1 | |
| A synthetic peptide corresponding to 17 amino acid peptide from 2nd extracellular domain of human TAAR1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only