missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabex-5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA544268
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Peptide sequence: MSLKSERRGI HVDQSDLLCK KGCGYYGNPA WQGFCSKCWR EEYHKARQKQ Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
RABGEF1 forms a complex with rabaptin-5 (RABPT5) that is required for endocytic membrane fusion, and it serves as a specific guanine nucleotide exchange factor (GEF) for RAB5.
Spécification
| Rabex-5 | |
| Polyclonal | |
| Unconjugated | |
| RABGEF1 | |
| BTB/POZ domain-containing protein KCTD7; RAB guanine nucleotide exchange factor (GEF) 1; RAB guanine nucleotide exchange factor 1; Rab5 exchange factor; rab5 GDP/GTP exchange factor; Rab5 GDP/GTP exchange factor, Rabex5; Rab5 guanine nucleotide exchange factor Rabex-5; Rab5ef; Rabaptin-5-associated exchange factor for Rab5; Rabex5; rabex-5; RABEX5 protein; RABGEF1; RAP1; Ras negative regulator Rabex-5; Ras negative regulator Rabex-5/Rin2; Rin2; zgc:65944 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27342 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UJ41 | |
| RABGEF1 | |
| Synthetic peptide directed towards the N-terminal of human RABGEF1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu