missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RACGAP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13739-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Rho GTPases control a variety of cellular processes. There are 3 subtypes of Rho GTPases in the Ras superfamily of small G proteins: RHO (see MIM 165370), RAC (see RAC1; MIM 602048), and CDC42 (MIM 116952). GTPase-activating proteins (GAPs) bind activated forms of Rho GTPases and stimulate GTP hydrolysis. Through this catalytic function, Rho GAPs negatively regulate Rho-mediated signals. GAPs may also serve as effector molecules and play a role in signaling downstream of Rho and other Ras-like GTPases.Spécification
RACGAP1 | |
Polyclonal | |
Unconjugated | |
RACGAP1 | |
HsCYK 4, ID GAP, KIAA1478, Male germ cell RacGap, MgcRacGAP, RACGAP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26934, 29127 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot, Immunoprecipitation | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H0H5, Q9WVM1 | |
RACGAP1 | |
RACGAP1 Fusion Protein Ag4676 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |