missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAI3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA533797
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Gibbon, Monkey, Marmoset (94%) Mouse, Rat, Hamster, Panda (88%) Bat, Dog (81%).
Retinoic acid plays a critical role in development, cellular growth, and differentiation. The specific function for this retinoic acid-induced gene has not yet been determined; however, it may play a role in embryonic development and epithelial cell differentiation. This gene is a member of the type 3 G protein-coupling receptor family, characterized by the signature 7-transmembrane domain motif. This gene may also be involved in interaction between retinoid acid and G protein signalling pathways. RAI3 has been reported in lung, kidney, colon, pancreas, prostate, and small intestine. ESTs have been isolated from amnion, breast, colon, eye, lung, nasal cavity, placenta, skin, uterus, and vessel libraries.
Spécification
| RAI3 | |
| Polyclonal | |
| Unconjugated | |
| GPRC5A | |
| G protein-coupled receptor class C group 5 member A; G protein-coupled receptor, class C, group 5, member A; G protein-coupled receptor, family C, group 5, member A; GPCR5A; Gprc5a; G-protein coupled receptor family C group 5 member A; orphan G-protein-coupling receptor PEIG-1; PEIG-1; Phorbol ester induced gene 1; phorbol ester induced protein-1; Rai3; Raig1; RAIG-1; retinoic acid induced 3; retinoic acid responsive; retinoic acid-induced gene 1 protein; Retinoic acid-induced protein 3; TIG1; TPA induced gene 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9052 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q8NFJ5 | |
| GPRC5A | |
| Synthetic 16 amino acid peptide from C-terminus of human GPRC5A / RAI3. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu