missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Raptor Recombinant Rabbit Monoclonal Antibody (1B10)

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA549939
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Raptor Recombinant Monoclonal Antibody for IHC (P), ELISA
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.
Spécification
| Raptor | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N122 | |
| RPTOR | |
| A synthesized peptide derived from human Raptor. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1B10 | |
| Unconjugated | |
| RPTOR | |
| 4932417H02Rik; KIAA1303; KOG1; Mip1; mKIAA1303; p150 target of rapamycin (TOR)-scaffold protein; p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats; Rap; RAPTOR; regulatory associated protein of MTOR complex 1; regulatory associated protein of MTOR, complex 1; regulatory-associated protein of mTOR; RGD1311784; RPTOR | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57521 | |
| -20°C or -80°C if preferred | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu