missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAR gamma-1 Recombinant Rabbit Monoclonal Antibody (PSH07-19)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Marque:  Invitrogen™ MA560185

Code nomenclature Nacres: NA.46

Code produit. 30484990

  • 441.00€ / 100µL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

Positive Control: SK-Br-3 cell lysate, T-47D cell lysate, A375 cell lysate, HeLa cell lysate, MEF cell lysate, Mouse skin tissue lysate, Rat skin tissue lysate, human skin tissue, mouse skin tissue, rat skin tissue. Tissue Specificity: Tissue enhanced (esophagus, skin). Subcellular Location: Nucleus, Cytoplasm. Sequence Similarities: 100% Mouse/Rat. Predicted band size: 50 kDa.

Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma. Two main isoforms of RAR gamma exist: RAR gamma-1 has been found to be predominantly expressed in skin, while RAR gamma-2 has been found early in embryogenesis and in embryonal carcinoma cells. RAR gamma is also known as RARD. The corresponding gene for the retinoic acid receptor gamma 1 is NR1B3.
TRUSTED_SUSTAINABILITY
Spécification

Spécification

RAR gamma-1
Recombinant Monoclonal
1 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P13631, P18911
Rarg
Recombinant protein within human RARG aa 155-454.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
PSH07-19
Unconjugated
Rarg
Nr1b3; Nuclear receptor subfamily 1 group B member 3; RAR gamma 2; RAR Gamma-1; RARC; RARD; Rarg; RAR-gamma; RARgamma2; retinoic acid; retinoic acid nuclear receptor gamma variant 1; retinoic acid nuclear receptor gamma variant 2; retinoic acid receptor; retinoic acid receptor gamma; retinoic acid receptor gamma 4; retinoic acid receptor, gamma
Rabbit
Protein A
RUO
19411, 5916, 685072
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis